Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl phosphonoacetate, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
867-13-0
C8H17O5P
224.19
GGUBFICZYGKNTD-UHFFFAOYSA-N
triethyl phosphonoacetate,ethyl 2-diethoxyphosphoryl acetate,ethyl diethoxyphosphoryl acetate,ethyl diethylphosphono acetate,triethyl phosphonoethanoate,diethyl carboethoxymethylphosphonate,diethyl ethoxycarbonylmethanephosphonate,diethylphosphonoacetic acid ethyl ester,diethyl carbethoxymethylphosphonate
13345
ethyl 2-diethoxyphosphorylacetate
CCOC(=O)CP(=O)(OCC)OCC
867-13-0
C8H17O5P
224.19
GGUBFICZYGKNTD-UHFFFAOYSA-N
triethyl phosphonoacetate,ethyl 2-diethoxyphosphoryl acetate,ethyl diethoxyphosphoryl acetate,ethyl diethylphosphono acetate,triethyl phosphonoethanoate,diethyl carboethoxymethylphosphonate,diethyl ethoxycarbonylmethanephosphonate,diethylphosphonoacetic acid ethyl ester,diethyl carbethoxymethylphosphonate
13345
ethyl 2-diethoxyphosphorylacetate
CCOC(=O)CP(=O)(OCC)OCC
for C-C Bond Formation (Olefination)
Specific Gravity 1.13
Melting Point -24.0°C
Refractive Index 1.4300 to 1.4320
Quantity 100g
Packaging Glass bottle
Density 1.1300g/mL
Boiling Point 260.0°C to 262.0°C
Color Undesignated
Flash Point 105°C
Infrared Spectrum Authentic
Acidity 2mg KOH/g max.
Assay Percent Range 97%
Linear Formula (C2H5O)2P(O)CH2CO2C2H5
Beilstein 04,I,573
Fieser 01,1216; 06,612; 12,527; 13,255
Solubility Information Solubility in water: insoluble
Formula Weight 224.19
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Triethyl phosphonoacetate
Show More Show Less