Learn More
Triethylamine, 99+%
Description
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizers for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in the automotive casting industry and the textile industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -115°C |
| Color | Colorless |
| Density | 0.726 g/mL |
| Boiling Point | 89°C to 90°C |
| Flash Point | -11°C (12°F) |
| Odor | Ammonia-like |
| Assay Percent Range | >99% |
| Refractive Index | 1.4005 |
| Linear Formula | (CH3CH2)3N |
| Quantity | 1 kg |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H311+H331-H314-H335-H336 |
| Hazard Statement | GHS H Statement H225-H314-H302-H312-H332 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIETHYLAMINE |
| EINECSNumber | 204-469-4 |
| RTECSNumber | YE0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.