Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, 99+%, Alfa Aesar™

Click to view available options
Quantity:
250g
1kg
3 x 1kg
121-44-8
C6H15N
101.193
MFCD00009051
ZMANZCXQSJIPKH-UHFFFAOYSA-N
triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane
8471
CHEBI:35026
N,N-diethylethanamine
CCN(CC)CC
121-44-8
C6H15N
101.193
MFCD00009051
ZMANZCXQSJIPKH-UHFFFAOYSA-N
triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane
8471
CHEBI:35026
N,N-diethylethanamine
CCN(CC)CC
Refractive Index 1.4005
Melting Point 115°C
Flash Point -11°C (12°F)
Density 0.726
Boiling Point 89°C to 90°C
Quantity 250g
Assay Percent Range >99%
Linear Formula (CH3CH2)3N
UN Number UN1296
Beilstein 1843166
Merck Index 14,9666
Solubility Information Miscible with water,ether and ethanol.
Formula Weight 101.19
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Triethylamine
Show More Show Less

GHS H Statement
H225-H314-H302-H312-H332

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H311+H331-H314-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIETHYLAMINE

EINECSNumber : 204-469-4

RTECSNumber : YE0175000

TSCA : Yes

Recommended Storage : Ambient temperatures