Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, 99%, pure, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
1L
2.5L
10L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
Plastic drum
121-44-8
C6H15N
101.19
ZMANZCXQSJIPKH-UHFFFAOYSA-N
triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane
8471
CHEBI:35026
N,N-diethylethanamine
CCN(CC)CC
121-44-8
C6H15N
101.19
ZMANZCXQSJIPKH-UHFFFAOYSA-N
triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane
8471
CHEBI:35026
N,N-diethylethanamine
CCN(CC)CC
Viscosity 0.36 mPa.s (20°C)
Packaging Glass bottle
Flash Point −11°C
Refractive Index 1.3990 to 1.4010
Infrared Spectrum Authentic
Quantity 5mL
Melting Point -115.0°C
Density 0.7280g/mL
Color Undesignated
Water 0.2% Max. (K.F.)
Boiling Point 90.0°C
Vapor Pressure 69mbar at 20°C
Specific Gravity 0.728
Assay Percent Range 98.5% min. (GC)
Linear Formula (C2H5)3N
Beilstein 04, 99
Fieser 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
Merck Index 15, 9831
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
Formula Weight 101.19
Physical Form Liquid
Percent Purity 99%
Grade Pure
Chemical Name or Material Triethylamine
Show More Show Less

GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Toxic in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

EINECSNumber : 204-469-4