Learn More
Triethylamine, 99%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -115.0°C |
| Density | 0.7280g/mL |
| Boiling Point | 90.0°C |
| Flash Point | −11°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Plastic drum |
| Molecular Formula | C6H15N |
| Refractive Index | 1.3990 to 1.4010 |
| Linear Formula | (C2H5)3N |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapour. Toxic if inhaled. Toxic in contact with skin. Harmful if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF IN EYES: Rinse cautiously wit |
| EINECSNumber | 204-469-4 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.