Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene glycol dimethyl ether, 99%, stabilized

Catalog No. p-3737628
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 112-49-2 | C8H18O4 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-49-2
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.23
MDL Number MFCD00008504
InChI Key YFNKIDBQEZZDLK-UHFFFAOYSA-N
Synonym triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
PubChem CID 8189
ChEBI CHEBI:44842
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
SMILES COCCOCCOCCOC

Specifications

Acidity 0.05mg KOH/g max.
Melting Point -45°C
Density 0.9860g/mL
Boiling Point 216°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4220 to 1.424
Linear Formula CH3OCH2CH2OCH2CH2OCH2CH2OCH3
Quantity 250 mL
Specific Gravity 0.986
Merck Index 15, 9861
Solubility Information Solubility in water: soluble. Other solubilities: miscible with hydrocarbon solvents
Stabilizer 0.01% BHT
Viscosity 2.5 mPa.s (20°C)
Formula Weight 178.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethylene glycol dimethyl ether, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
May form explosive peroxides.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if p
EINECSNumber 203-977-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.