Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol, Purified, 97%, Spectrum™ Chemical
SDP

Catalog No. 18614551 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can

Catalog No. 18-614-551

Supplier: Spectrum Chemical Manufacturing Corporation T111820LTMT

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
SMILES OCCOCCOCCO

Specifications

CAS Max % 1
Packaging Steel Can
Quantity 20 L
Percent Purity 0.97
Grade Purified

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.