Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoromethanesulfonamide, 96%

Catalog No. p-7026223
Click to view available options
Quantity:
1 g
5 g

CAS: 421-85-2 | CH2F3NO2S | 149.09 g/mol

Trifluoromethanesulfonamide is used as an intermediate in synthetic chemistry. It is used in the preparation of specific inhibitors of mammalian secreted phospholipases A2. It is also involved in the synthesis of ecicosanoids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoromethanesulfonamide is used as an intermediate in synthetic chemistry. It is used in the preparation of specific inhibitors of mammalian secreted phospholipases A2. It is also involved in the synthesis of ecicosanoids.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-85-2
Molecular Formula CH2F3NO2S
Molecular Weight (g/mol) 149.09
MDL Number MFCD00068714
InChI Key KAKQVSNHTBLJCH-UHFFFAOYSA-N
Synonym trifluoromethanesulphonamide, trifluoromethane sulfonamide, trifluoromethylsulfonamide, 1,1,1-trifluoromethanesulfonamide, chembl67511, methanesulfonamide, 1,1,1-trifluoro, 1bcd, trifluoromethyl sulfonamide, triflamide, trifluorometanesulfonamide
PubChem CID 79001
IUPAC Name trifluoromethanesulfonamide
SMILES NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 116°C to 119°C
Quantity 1 g
Beilstein 1812099
Solubility Information Soluble in water.
Formula Weight 149.09
Percent Purity 96%
Chemical Name or Material Trifluoromethanesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 431-270-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.