Learn More
Trifluoromethanesulfonic anhydride, 98%
Description
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.
Solubility
Miscible with dichloromethane. Immiscible with hydrocarbons.
Notes
Moisture sensitive. Incompatible with water, strong bases, strong oxidizing agents, acids and alcohols.
Specifications
Specifications
| Melting Point | -80°C |
| Density | 1.72 |
| Boiling Point | 81°C to 83°C |
| Refractive Index | 1.321 |
| Linear Formula | (CF3SO2)2O |
| Quantity | 10 g |
| UN Number | UN3265 |
| Beilstein | 1813600 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Miscible with dichloromethane. Immiscible with hydrocarbons. |
| Show More |
Safety and Handling
| Hazard Category | H290-H302-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: I |
| EINECSNumber | 206-616-8 |
| RTECSNumber | PB2772000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.