Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM Antibody (TRIM-04), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB50485A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50485A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50485A532 Supplier Novus Biologicals Supplier No. NB500485AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIM Monoclonal specifically detects TRIM in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen TRIM
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone TRIM-04
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062
Gene Symbols TRAT1
Host Species Mouse
Immunogen Recombinant fragment, corresponding to amino acids 29-186 of Human TRIM.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 50852.0
Test Specificity The antibody TRIM-04 reacts with T cell receptor-interacting molecule (TRIM), a 30 kDa adaptor protein expressed by T cells.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.