Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220704 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220704 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220704 Supplier Novus Biologicals Supplier No. NBP220704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM10 Polyclonal specifically detects TRIM10 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TRIM10
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.98mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UDY6
Gene Alias B30-RING finger protein, HERF1hematopoietic RING finger 1, RFB30MGC141979, RING finger protein 9, RNF9tripartite motif protein 10, tripartite motif containing 10, tripartite motif-containing 10, tripartite motif-containing protein 10, Zn-finger protein
Gene Symbols TRIM10
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TRIM10. The exact sequence is proprietary.
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10107
Test Specificity Human. Species reactivity based on sequence identity: Monkey (95%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.