Learn More
Description
Specifications
Specifications
| Antigen | TRIM17 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | NP_001020111 |
| Gene Alias | E3 ubiquitin-protein ligase TRIM17, EC 6.3.2.-, RBCCRNF16terf, RING finger protein 16, RING finger protein terf, TERF, Testis RING finger protein, tripartite motif containing 17, tripartite motif-containing 17, Tripartite motif-containing protein 17 |
| Gene Symbols | TRIM17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
