Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM21/SSA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303809100UL
Description
TRIM21/SSA1 Polyclonal specifically detects TRIM21/SSA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| TRIM21/SSA1 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| 52 kDa ribonucleoprotein autoantigen Ro/SS-A, 52 kDa Ro/SSA, E3 ubiquitin-protein ligase TRIM21, EC 6.3.2.-, rhTRIM21, RNF81RING finger protein 81, ro(SS-A), RO52SS-A, Sicca syndrome antigen A, Sjoegren syndrome type A antigen, Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro), SSA1SSA, tripartite motif containing 21,52 kDa Ro protein, tripartite motif-containing 21, Tripartite motif-containing protein 21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-330 of human TRIM21/SSA1 (NP_003132.2). | |
| 100 μL | |
| Zinc Finger | |
| 6737 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction