Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trim22 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094501 20 μL
NB094500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094501 Supplier Novus Biologicals Supplier No. NBP30485520UL

Rabbit Polyclonal Antibody

Trim22 Polyclonal specifically detects Trim22 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Trim22
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 50 kDa-stimulated trans-acting factor, E3 ubiquitin-protein ligase TRIM22, EC 6.3.2, EC 6.3.2.-, GPSTAF50, RING finger protein 94, RNF94stimulated trans-acting factor (50 kDa), Staf-50, STAF50staf-50, tripartite binding motif 22, tripartite motif containing 22, tripartite motif protein TRIM22, tripartite motif-containing 22, Tripartite motif-containing protein 22
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 215-411 of human TRIM22 (NP_006065.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10346
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.