Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM25 Antibody (5B5B12) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261814 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61814 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61814 Supplier Novus Biologicals Supplier No. NBP261814
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIM25 Monoclonal specifically detects TRIM25 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TRIM25
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5B5B12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 6.3.2.19, EC 6.3.2.n3, EFPZ147, Estrogen-responsive finger protein, RING finger protein 147, RNF147E3 ubiquitin/ISG15 ligase TRIM25, tripartite motif containing 25, tripartite motif-containing 25, Tripartite motif-containing protein 25, Ubiquitin/ISG15-conjugating enzyme TRIM25, Zinc finger protein 147, zinc finger protein 147 (estrogen-responsive finger protein), zinc finger protein-147, ZNF147tripartite motif protein TRIM25
Gene Symbols TRIM25
Host Species Mouse
Immunogen Purified recombinant fragment of human TRIM25 (AA: 211-360) expressed in E. Coli.
Molecular Weight of Antigen 68.7 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7706
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.