Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM25 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB194338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB194338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB194338 Supplier Novus Biologicals Supplier No. NBP176826G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM25 Polyclonal antibody specifically detects TRIM25 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen TRIM25
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.19, EC 6.3.2.n3, EFPZ147, Estrogen-responsive finger protein, RING finger protein 147, RNF147E3 ubiquitin/ISG15 ligase TRIM25, tripartite motif containing 25, tripartite motif-containing 25, Tripartite motif-containing protein 25, Ubiquitin/ISG15-conjugating enzyme TRIM25, Zinc finger protein 147, zinc finger protein 147 (estrogen-responsive finger protein), zinc finger protein-147, ZNF147tripartite motif protein TRIM25
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human TRIM25. The immunogen is located within amino acids 40 - 90 of TRIM25. Amino Acid Squence: CRAVYQARPQLHKNT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7706
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.