Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM29 Antibody, Novus Biologicals™
SDP

Catalog No. NB12022207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12022207 0.1 mg
NB12022207T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB12022207 Supplier Novus Biologicals Supplier No. NB12022207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

TRIM29 Polyclonal specifically detects TRIM29 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen TRIM29
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. Q14134
Gene Alias Ataxia telangiectasia group D-associated protein, ATDCataxia-telangiectasia group D-associated protein, FLJ36085, tripartite motif containing 29, tripartite motif protein TRIM29, tripartite motif-containing 29, tripartite motif-containing protein 29
Gene Symbols TRIM29
Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 569-583 of human TRIM29.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23650
Test Specificity Reacts with Human. Predicted to react with Mouse, Rat, Chimpanzee and Canine.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.