Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM34 Antibody, Novus Biologicals™
SDP

Catalog No. NBP18035920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18035920 20 μL
NBP180359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18035920 Supplier Novus Biologicals Supplier No. NBP18035920UL

Rabbit Polyclonal Antibody

TRIM34 Polyclonal specifically detects TRIM34 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRIM34
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_569074
Gene Alias EC 6.3.2, interferon-responsive, Interferon-responsive finger protein 1, RING finger protein 21, RNF21tripartite motif-containing protein 34, tripartite motif containing 34, tripartite motif-containing 34
Gene Symbols TRIM34
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human TRIM34. Peptide sequence CRACITVSNKEAVTSMGGKSSCPVCGISYSFEHLQANQHLANIVERLKEV.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 53840
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.