Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM34 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | TRIM34 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18035920
![]() |
Novus Biologicals
NBP18035920UL |
20 μL |
Each for $158.00
|
|
|||||
NBP180359
![]() |
Novus Biologicals
NBP180359 |
100 μL |
Each for $487.50
|
|
|||||
Description
TRIM34 Polyclonal specifically detects TRIM34 in Human samples. It is validated for Western Blot.Specifications
TRIM34 | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
NP_569074 | |
53840 | |
Synthetic peptide directed towards the N terminal of human TRIM34. Peptide sequence CRACITVSNKEAVTSMGGKSSCPVCGISYSFEHLQANQHLANIVERLKEV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
EC 6.3.2, interferon-responsive, Interferon-responsive finger protein 1, RING finger protein 21, RNF21tripartite motif-containing protein 34, tripartite motif containing 34, tripartite motif-containing 34 | |
TRIM34 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title