Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM34 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen TRIM34
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP18005020
SDP
View Documents
Novus Biologicals
NBP18005020UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP180050
SDP
View Documents
Novus Biologicals
NBP180050
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

TRIM34 Polyclonal specifically detects TRIM34 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TRIM34
Polyclonal
Rabbit
Zinc Finger
EC 6.3.2, interferon-responsive, Interferon-responsive finger protein 1, RING finger protein 21, RNF21tripartite motif-containing protein 34, tripartite motif containing 34, tripartite motif-containing 34
TRIM34
IgG
31 kDa
Western Blot
Unconjugated
RUO
NP_569074
53840
Synthetic peptide directed towards the N terminal of human TRIM34. Peptide sequence SMGGKSSCPVCGISYSFEHLQANQHLANIVERLKEVKLSPDNGKKRDLCD.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.