Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM37 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB106073 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB106073 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB106073 Supplier Novus Biologicals Supplier No. NBP298831UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM37 Polyclonal specifically detects TRIM37 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TRIM37
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias EC 6.3.2.-, KIAA0898Tripartite motif-containing protein 37, Mulibrey nanism protein, MULRING-B-box-coiled-coil protein, POB1E3 ubiquitin-protein ligase TRIM37, TEF3, tripartite motif containing 37, tripartite motif-containing 37
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human TRIM37.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4591
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.