Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM38 Antibody (OTI113D3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274610FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274610FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274610FR Supplier Novus Biologicals Supplier No. NBP274610FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIM38 Monoclonal specifically detects TRIM38 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TRIM38
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI113D3
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias MGC8946, RING finger protein 15RORETRo/SSA ribonucleoprotein homolog, RNF15, tripartite motif containing 38, tripartite motif-containing 38, tripartite motif-containing protein 38, Zinc finger protein RoRet, zinc-finger protein RoRet
Gene Symbols TRIM38
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-265 of human TRIM38 (NP_006346) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10475.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.