Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM41 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB335290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335290 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335290 Supplier Novus Biologicals Supplier No. NBP305293MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM41 Polyclonal antibody specifically detects TRIM41 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TRIM41
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias tripartite motif containing 41
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the human TRIM41 protein (between amino acids 400-470) [UniProt Q8WV44].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 90933
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.