Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM42 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220718 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220718 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220718 Supplier Novus Biologicals Supplier No. NBP220718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM42 Polyclonal specifically detects TRIM42 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRIM42
Applications Western Blot
Classification Polyclonal
Concentration 0.58mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Gene Alias FLJ40097, MGC163346, tripartite motif containing 42, tripartite motif-containing 42, tripartite motif-containing protein 42
Gene Symbols TRIM42
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TRIM42. The exact sequence is proprietary.
Molecular Weight of Antigen 83 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287015
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.