Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM47 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 268851AP150UL

Description
Synaptogyrins comprise a family of tyrosine-phosphorylated proteins with two neuronal (synaptogyrins 1 and 3) isoforms. Previous studies have indicated that synaptogyrins are involved in the regulation of neurotransmitter release. Synaptogyrin 1 is an integral membrane protein ubiquitously expressed in the nervous system. Synaptogyrin has four transmembrane domains and an extended cytoplasmic tail but it is not glycosylated. Synaptogyrin 1 is a tyrosine-phosphorylated by c-src and possibly other tyrosine kinases. The function of Synaptogyrin 1 is not well understood, but it may be a useful marker for nerve terminals and diagnosing neuroendocrine tumors.Specifications
TRIM47 | |
Polyclonal | |
Unconjugated | |
TRIM47 | |
GOA, RING finger protein 100, RNF100, TRIM47, tripartite motif containing 47 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
91107 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96LD4 | |
TRIM47 | |
TRIM47 Fusion Protein Ag22868 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction