Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM47 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TRIM47 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5487
![]() ![]() |
Proteintech Group Inc
268851AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Synaptogyrins comprise a family of tyrosine-phosphorylated proteins with two neuronal (synaptogyrins 1 and 3) isoforms. Previous studies have indicated that synaptogyrins are involved in the regulation of neurotransmitter release. Synaptogyrin 1 is an integral membrane protein ubiquitously expressed in the nervous system. Synaptogyrin has four transmembrane domains and an extended cytoplasmic tail but it is not glycosylated. Synaptogyrin 1 is a tyrosine-phosphorylated by c-src and possibly other tyrosine kinases. The function of Synaptogyrin 1 is not well understood, but it may be a useful marker for nerve terminals and diagnosing neuroendocrine tumors.Specifications
TRIM47 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96LD4 | |
91107 | |
TRIM47 Fusion Protein Ag22868 | |
Primary | |
-20°C | |
TRIM47 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GOA, RING finger protein 100, RNF100, TRIM47, tripartite motif containing 47 | |
TRIM47 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title