Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM5 alpha Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB215268 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB215268 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB215268 Supplier Novus Biologicals Supplier No. NBP177207JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM5 alpha Polyclonal antibody specifically detects TRIM5 alpha in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TRIM5 alpha
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 19 amino acids near the center of rhesus monkey TRIM5 alpha. The immunogen is located within amino acids 390 - 440 of TRIM5 alpha. Amino Acid Squence: GLQEGVKYSVFQDG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Lipid and Metabolism, Virology Bacteria and Parasites, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 85363
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.