Learn More
Description
Specifications
Specifications
| Antigen | TRIM56 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 |
| Formulation | PBS & 2% Sucrose. with No Preservative |
| Gene Accession No. | NP_112223 |
| Gene Alias | DKFZp667O116, E3 ubiquitin-protein ligase TRIM56, EC 6.3.2.-, RING finger protein 109, RNF109FLJ35608, tripartite motif containing 56, tripartite motif-containing 56, Tripartite motif-containing protein 56 |
| Gene Symbols | TRIM56 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
