Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM56 Antibody (OTI5A1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N274613A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274613A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274613A647 Supplier Novus Biologicals Supplier No. NBP274613AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIM56 Monoclonal specifically detects TRIM56 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TRIM56
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5A1
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp667O116, E3 ubiquitin-protein ligase TRIM56, EC 6.3.2.-, RING finger protein 109, RNF109FLJ35608, tripartite motif containing 56, tripartite motif-containing 56, Tripartite motif-containing protein 56
Gene Symbols TRIM56
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 311-554 of human TRIM56 (NP_112223) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 81844.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.