Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM58 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017181 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017181 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017181 Supplier Novus Biologicals Supplier No. NBP281870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM58 Polyclonal specifically detects TRIM58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TRIM58
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, ELISA
Formulation PBS
Gene Alias BIA2, DKFZp434C091, Protein BIA2, tripartite motif containing 58, tripartite motif-containing 58, tripartite motif-containing protein 58
Gene Symbols TRIM58
Host Species Rabbit
Immunogen TRIM58 antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TRIM58. Amino Acid Squence: DTLPRKGETTPSPENGV
Molecular Weight of Antigen 56 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 25893
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.