Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM58 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342065 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342065 Supplier Novus Biologicals Supplier No. NBP281870MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM58 Polyclonal antibody specifically detects TRIM58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen TRIM58
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias BIA2, DKFZp434C091, Protein BIA2, tripartite motif containing 58, tripartite motif-containing 58, tripartite motif-containing protein 58
Host Species Rabbit
Immunogen TRIM58 antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TRIM58. Amino Acid Squence: DTLPRKGETTPSPENGV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 25893
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.