Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM59 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB185486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185486 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185486 Supplier Novus Biologicals Supplier No. NBP300528UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIM59 Polyclonal antibody specifically detects TRIM59 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TRIM59
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MGC129860, MGC129861, Mrf1, RING finger protein 104, RNF104MGC26631, TRIM57, tripartite motif containing 59, tripartite motif-containing 57, tripartite motif-containing 59, tripartite motif-containing protein 59, TSBF1MRF1, tumor suppressor TSBF1, Tumor suppressor TSBF-1
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the human TRIM59 protein (between amino acids 250-300) [UniProt Q8IWR1]
Purification Method Immunogen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 286827
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.