Learn More
Description
Specifications
Specifications
| Antigen | GERP |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal |
| Gene Accession No. | Q9BZR9 |
| Gene Alias | EC 6.3.2.-, GERPRING finger protein 27glioblastoma expressed ring finger protein, Glioblastoma-expressed RING finger protein, RNF27probable E3 ubiquitin-protein ligase TRIM8, tripartite motif containing 8, tripartite motif protein TRIM8, tripartite motif-containing 8, Tripartite motif-containing protein 8 |
| Gene Symbols | TRIM8 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
