Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM8 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001192 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001192 Supplier Novus Biologicals Supplier No. NB1001192
Add to Cart
Add to Cart

Goat Polyclonal Antibody

GERP Polyclonal specifically detects GERP in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA.

Specifications

Antigen GERP
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 4 - 6 ug/ml, Immunohistochemistry-Paraffin 4 - 6 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_112174.
Gene Alias EC 6.3.2.-, GERPRING finger protein 27glioblastoma expressed ring finger protein, Glioblastoma-expressed RING finger protein, RNF27probable E3 ubiquitin-protein ligase TRIM8, tripartite motif containing 8, tripartite motif protein TRIM8, tripartite motif-containing 8, Tripartite motif-containing protein 8
Gene Symbols TRIM8
Host Species Goat
Immunogen Peptide with sequence C-YGQPSTKHYVTS corresponding to C-Terminus according to NP_112174.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 81603
Test Specificity Human. Expected to react based on sequence similarity: Mouse, Rat, Dog, Pig.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.