Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM9 Antibody (OTI2A1), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB282297 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB282297 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB282297 Supplier Novus Biologicals Supplier No. NBP274618MFV500

Mouse Monoclonal Antibody

TRIM9 Monoclonal antibody specifically detects TRIM9 in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen TRIM9
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2A1
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.-, KIAA0282homolog of rat RING finger Spring, RNF91RING finger protein 91, SPRINGE3 ubiquitin-protein ligase TRIM9, tripartite motif containing 9, tripartite motif-containing 9, Tripartite motif-containing protein 9
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 284-669 of human TRIM9 (NP_443210) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 114088
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.