Learn More
Trimethyl orthoacetate, 98%
Description
Trimethyl orthoacetate is used as a diols can be protected as their 1-methoxyethylidene cyclic acetals (2-methoxy-2-methyl-1,3-dioxolanes) by acid-catalyzed exchange. It is mainly used as the chemical intermediates of the medicine. Some of the other applications includes it is mainly used as chemical intermediate of pharmaceutical and agricultural chemicals, used for compound medical intermediates of vitamin B1, vitamin A1 and sulfanilamide organism, used for dye and spices industry, Medicine, agricultural chemicals, paint additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trimethyl orthoacetate is used as a diols can be protected as their 1-methoxyethylidene cyclic acetals (2-methoxy-2-methyl-1,3-dioxolanes) by acid-catalyzed exchange. It is mainly used as the chemical intermediates of the medicine. Some of the other applications includes it is mainly used as chemical intermediate of pharmaceutical and agricultural chemicals, used for compound medical intermediates of vitamin B1, vitamin A1 and sulfanilamide organism, used for dye and spices industry, Medicine, agricultural chemicals, paint additives.
Solubility
It hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
| Density | 0.956 |
| Boiling Point | 107°C to 109°C |
| Flash Point | 16°C (60°F) |
| Refractive Index | 1.389 |
| Linear Formula | CH3C(OCH3)3 |
| Quantity | 50 mL |
| UN Number | UN3272 |
| Beilstein | 1098338 |
| Sensitivity | Moisture sensitive |
| Solubility Information | It hydrolyzes in water. |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H317-H319 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P303+P361+P353-P305+P351+P338-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S. |
| EINECSNumber | 215-892-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.