Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl orthopropionate, 97%, Acros Organics

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
24823-81-2
C6H14O3
134.18
MFCD00043305
ZGMNAIODRDOMEK-UHFFFAOYSA-N
trimethyl orthopropionate,propane, 1,1,1-trimethoxy,trimethylorthopropionate,trimethoxypropane,acmc-209gew,trimethyl orthopropanoate,propane,1,1,1-trimethoxy,ksc489s6b,zgmnaiodrdomek-uhfffaoysa,orthopropionic acid trimethyl ester
141145
1,1,1-trimethoxypropane
CCC(OC)(OC)OC
24823-81-2
C6H14O3
134.18
MFCD00043305
ZGMNAIODRDOMEK-UHFFFAOYSA-N
trimethyl orthopropionate,propane, 1,1,1-trimethoxy,trimethylorthopropionate,trimethoxypropane,acmc-209gew,trimethyl orthopropanoate,propane,1,1,1-trimethoxy,ksc489s6b,zgmnaiodrdomek-uhfffaoysa,orthopropionic acid trimethyl ester
141145
1,1,1-trimethoxypropane
CCC(OC)(OC)OC
Boiling Point 121°C to 122°C
Flash Point 19°C
Specific Gravity 0.944
Refractive Index 1.3970 to 1.399
Density 0.9440g/mL
Color Colorless
Infrared Spectrum Authentic
Quantity 25mL
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula CH3CH2C(OCH3)3
Formula Weight 134.18
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Trimethyl orthopropionate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger