Learn More
Trimethylgermanium chloride
Description
Trimethylgermanium chloride is used to prepare trimethyl(2-thienyl)germane. It is also used as alkylating and precursor to prepare Grignard reagent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -14°C |
| Density | 1.249 g/mL |
| Boiling Point | 102°C |
| Flash Point | 1°C (35°F) |
| Refractive Index | 1.4337 |
| Quantity | 1 g |
| UN Number | UN2924 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Immiscible with water.Miscible with ether,tetrahydrofuran,hexane,benzene,dichloromethane and chloroform. |
| Formula Weight | 153.15 |
| Show More |
Safety and Handling
| Hazard Category | H225-H314 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| EINECSNumber | 216-214-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.