Learn More
Trimethyloxonium tetrafluoroborate, 96%
Description
Trimethyloxonium tetrafluoroborate act as a avmethylating agent and activates C-X multiple bonds. It is involved in the esterification of polyfunctional carboxylic acids. It acts as a catalyst in the polymerization of cyclic sulfides and ethers. Further, it is used in Beckmann rearrangement of oximes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trimethyloxonium tetrafluoroborate act as a avmethylating agent and activates C-X multiple bonds. It is involved in the esterification of polyfunctional carboxylic acids. It acts as a catalyst in the polymerization of cyclic sulfides and ethers. Further, it is used in Beckmann rearrangement of oximes.
Solubility
Soluble in nitrobenzene, nitromethane, chloroform, hot acetone, liquid sulfur dioxide. Slightly soluble in dichloromethane. Insoluble in common organic solvents.
Notes
Hygroscopic. Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.
Specifications
Specifications
| Melting Point | ∽190°C (decomposition) |
| Assay Percent Range | 96% |
| Quantity | 10 g |
| Beilstein | 3597303 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in nitrobenzene,nitromethane,chloroform,hot acetone,liquid sulfur dioxide. Slightly soluble in dichloromethane. Insoluble in common organic solvents. |
| IUPAC Name | trimethyloxidanium;tetrafluoroborate |
| Formula Weight | 147.91 |
| Chemical Name or Material | Trimethyloxonium Tetrafluoroborate |
Safety and Handling
| Hazard Category | H314-H335 |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 206-994-4 |
| TSCA | Yes |
| storageNote1 | Moisture sensitive |
| Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.