Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilanol

Catalog No. p-7077740
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-40-6
Molecular Formula C3H9LiOSi
Molecular Weight (g/mol) 96.13
MDL Number MFCD02751657
InChI Key OXOZHAWWRPCVGL-UHFFFAOYSA-N
Synonym trimethylsilanol, silanol, trimethyl, unii-z4bin3300p, trimethylhydroxysilane, ccris 1320, hydroxy trimethyl silane, silanol, 1,1,1-trimethyl, ksc504m9b, 2004-14-0 lithium salt, 10519-96-7 potassium salt
PubChem CID 66110
IUPAC Name hydroxy(trimethyl)silane
SMILES [Li+].C[Si](C)(C)[O-]

Specifications

Melting Point -12.0°C
Density 0.8140g/mL
Boiling Point 98.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Linear Formula (CH3)3SiOH
Quantity 5 g
Specific Gravity 0.814
Solubility Information Solubility in water: 0.995g/L.
Viscosity 4 mPas (25°C)
Formula Weight 90.2
Percent Purity ≥97.5% (GC)
Chemical Name or Material Trimethylsilanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 213-914-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.