Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trimethylsilyl)methyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. T157925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T157925G 25 g
T15791G 1 g
T15795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T157925G Supplier TCI America Supplier No. T157925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64035-64-9
Molecular Formula C5H11F3O3SSi
Molecular Weight (g/mol) 236.28
MDL Number MFCD00009914
InChI Key VMDMAAJZSXXCQV-UHFFFAOYSA-N
Synonym trimethylsilyl methyl trifluoromethanesulfonate, trimethylsilylmethyl triflate, trimethylsilyl methyl triflate, trimethylsilylmethyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, trimethylsilyl methyl ester, acmc-1b9lz, trimethylsiylmethyltrifluoromethanesulfnate, trimethylsilylmethyltrifluoromethanesulfonate, trifluoromethanesulfonic acid trimethylsilylmethyl ester
PubChem CID 2760857
IUPAC Name trimethylsilylmethyl trifluoromethanesulfonate
SMILES C[Si](C)(C)COS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 168°C
Quantity 25 g
UN Number 2920
Formula Weight 236.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Trimethylsilyl)methyl Trifluoromethanesulfonate [Trimethylsilylmethylating Reagent]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.