Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl trifluoromethanesulfonate, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
27607-77-8
C4H9F3O3SSi
222.26
MFCD00000406
FTVLMFQEYACZNP-UHFFFAOYSA-N
trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate
65367
trimethylsilyl trifluoromethanesulfonate
C[Si](C)(C)OS(=O)(=O)C(F)(F)F
27607-77-8
C4H9F3O3SSi
222.26
MFCD00000406
FTVLMFQEYACZNP-UHFFFAOYSA-N
trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate
65367
trimethylsilyl trifluoromethanesulfonate
C[Si](C)(C)OS(=O)(=O)C(F)(F)F
Boiling Point 140°C
Specific Gravity 1.225
Flash Point 25°C
Refractive Index 1.3600 to 1.362
Density 1.2250g/mL
Infrared Spectrum Authentic
Quantity 10mL
Packaging Glass bottle
Color Colorless to Brown
Assay Percent Range 99%
Linear Formula CF3SO3Si(CH3)3
Fieser 08,514; 09,497; 10,438; 11,584; 12,543; 13,118; 14,333; 16,363; 17,379
Merck Index 15, 9891
Solubility Information Solubility in water: reacts
Formula Weight 222.26
Physical Form Fuming Liquid
Percent Purity 99%
Chemical Name or Material Trimethylsilyl trifluoromethanesulfonate
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 248-565-4