Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIML1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP154340 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP154340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP154340 Supplier Novus Biologicals Supplier No. NBP154340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIML1 Polyclonal specifically detects TRIML1 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRIML1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8N9V2
Gene Alias EC 6.3.2.-, FLJ36180, MGC138638, probable E3 ubiquitin-protein ligase TRIML1, RING finger protein 209, RNF209MGC138639, tripartite motif family-like 1, Tripartite motif family-like protein 1
Gene Symbols TRIML1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TRIML1(tripartite motif family-like 1) The peptide sequence was selected from the middle region of TRIML1. Peptide sequence DSVSRKGNLPKPPGDLFSLIGLKIGDDYSLWVSSPLKGQHVREPVCKVGV.
Molecular Weight of Antigen 53 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 339976
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.