Learn More
Invitrogen™ TRIO Polyclonal Antibody

Description
TRIO Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | TRIO |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | triob |
| Gene Accession No. | Q1LUA6 |
| Gene Alias | 6720464I07Rik; AA408740; ARHGEF23; LOW QUALITY PROTEIN: triple functional domain protein; PTPRF-interacting protein; si:ch211-13c6.1; si:dkey-158b13.2; Solo; tgat; trio; trio Rho guanine nucleotide exchange factor; trio Rho guanine nucleotide exchange factor a; trioa; triple functional domain (PTPRF interacting); triple functional domain protein |
| Gene Symbols | trioa, triob |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.