Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triosephosphate isomerase Antibody (181), Novus Biologicals™
SDP

Catalog No. NBP243761 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243761 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243761 Supplier Novus Biologicals Supplier No. NBP243761
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence

Triosephosphate isomerase Monoclonal specifically detects Triosephosphate isomerase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Triosephosphate isomerase
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 181
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS with No Preservative
Gene Alias EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1
Gene Symbols TPI1
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Triosephosphate isomerase. The exact sequence is proprietary.
Molecular Weight of Antigen 31 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7167
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.