Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIP6 Antibody, Novus Biologicals™
SDP

Catalog No. NB10079791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10079791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10079791 Supplier Novus Biologicals Supplier No. NB10079791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

TRIP6 Polyclonal specifically detects TRIP6 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRIP6
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_003293.1
Gene Alias MGC10556, MGC10558, MGC29959, MGC3837, MGC4423, OIP1OIP-1, Opa-interacting protein 1, thyroid hormone receptor interactor 6, thyroid receptor-interacting protein 6, TR-interacting protein 6, ZRP-1TRIP-6, zyxin related protein 1, Zyxin-related protein 1
Gene Symbols TRIP6
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human Thyroid Hormone Receptor Interactor 6 using the numbering given in entry NP_003293.1 (GeneID 7205).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7205
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.