Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringe
Catalog No. p-7139600
Click to view available options
Volume (Metric):
100 μL
250 μL
Needle Gauge:
23 G
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Needle), Stainless Steel (Plunger)
Description 250 μL FN GT, 23 G, 57 mm
Needle Gauge 23 G
Length (Metric) 57 mm
Product Type GC Autosampler Syringe
Quantity Ea.
Volume (Metric) 250 μL
Length (Metric) Needle 57 mm
Needle Point Style Side Hole
For Use With (Equipment) TriPlus RSH Autosampler
Connection Type Fixed
Sterility Sterile
PTFE Tipped Plunger Yes
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.