Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIS-Buffered Saline (TBS, 20X), With 2% Tween 20, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2 L
Description
Applications
TRIS-buffered saline (TBS, 20X), with 2% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein:protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.
Solubility
It is soluble in water.
Notes
1.0 M Tris-HCl, 3.0 M NaCl, pH 7.4 ± 0.1 with 2.0% Tween-20

Specifications
Specifications
Product Type | TBS |
Quantity | 2 L |
Form | Liquid |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction