Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(isopropylcyclopentadienyl)lanthanum(III), 99.9% (REO)

Catalog No. AA3932303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3932303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3932303 Supplier Thermo Scientific Chemicals Supplier No. 03932303
Only null left
Add to Cart

CAS: 68959-87-5 | C24H33La | 460.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68959-87-5
Molecular Formula C24H33La
Molecular Weight (g/mol) 460.43
MDL Number MFCD00145482
InChI Key BHWFVSOAOOHARF-UHFFFAOYNA-N
Synonym lanthanum,tris 1,2,3,4,5-h-1-1-methylethyl-2,4-cyclopentadien-1-yl

Specifications

Color Colorless to Yellow
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 460.43
Percent Purity 99.9%
Physical Form Liquid
Assay (REO)
Chemical Name or Material Tris(isopropylcyclopentadienyl)lanthanum(III)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.