Learn More
Triton X-100
Description
Detergent, equivalent to Nonidet P-40.Triton™ X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Detergent, equivalent to Nonidet P-40.Triton|r X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
Specifications
Specifications
| Boiling Point | >250°C |
| Melting Point | ∽5°C |
| Density | 1.070 |
| Quantity | 2500 mL |
| Flash Point | 180°C (356°F) |
| Refractive Index | 1.49 |
| Solubility Information | Miscible with water. |
| IUPAC Name | 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol |
| Formula Weight | ca. 625 |
| Chemical Name or Material | Triton X-100 |
Safety and Handling
| Hazard Category | H302-H318 |
| Precautionary Statement | P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c |
| RTECSNumber | MD0907600 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.